Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(1,2-Dimethyl-3-indolyl)-3-[4-(diethylamino)-2-methylphenyl]phthalide 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A210353-10G A210353-1G A210353-25G A210353-5G
77012-930EA 32 USD InStock
77012-930 77012-932 77012-934 77683-596
3-(1,2-Dimethyl-3-indolyl)-3-[4-(diethylamino)-2-methylphenyl]phthalide 95%, Ambeed.Inc
3-(1,2-Dimethyl-3-indolyl)-3-[4-(diethylamino)-2-methylphenyl]phthalide
Precaución: For research use only.


Formula: C₂₉H₃₀N₂O₂
MDL Number: MFCD08276348
CAS Number: 36499-49-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty