Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Hydroxy-3'-methylacetophenone 98%

Supplier: AMBEED, INC
Warning

A211522-10G A211522-1G A211522-25G A211522-5G A211522-100G
77030-520EA 27 USD InStock
77030-520 77030-522 77030-524 77030-526 77030-528 77671-888
4'-Hydroxy-3'-methylacetophenone 98%
4'-Hydroxy-3'-methylacetophenone
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD00002231
CAS Number: 876-02-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty