Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyanophenyl-4-butylbenzoate 97%

Supplier: AMBEED, INC
Warning

A212184-10G A212184-25G A212184-5G
77030-548EA 26 USD InStock
77030-548 77030-550 77671-892
4-Cyanophenyl-4-butylbenzoate 97%
4-Cyanophenyl 4-butylbenzoate
Precaución: for Research Use Only


Formula: C₁₈H₁₇NO₂
MDL Number: MFCD00001816
CAS Number: 38690-77-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty