Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromodibenzofuran 97%

Supplier: AMBEED, INC
Warning

A212767-100G A212767-100MG A212767-1G A212767-250MG A212767-5G
76986-770EA 675 USD InStock
76986-770 76986-772 76986-774 76986-776 76986-778
1-Bromodibenzofuran 97%
1-Bromodibenzofuran
Precaución: for Research Use Only


Formula: C₁₂H₇BrO
MDL Number: MFCD00094238
CAS Number: 50548-45-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty