Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Aspartic acid 4-cyclohexyl ester 98%

Supplier: AMBEED, INC
Warning

A213441-1G A213441-250MG A213441-5G
76999-670EA 10 USD InStock
76999-670 76999-672 76999-674
L-Aspartic acid 4-cyclohexyl ester 98%
L-Aspartic acid 4-cyclohexyl ester
Precaución: for Research Use Only


Formula: C₁₀H₁₇NO₄
MW: 215.249 g/mol
MDL Number: MFCD00190762
CAS Number: 112259-66-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty