Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-(bromomethyl)-2-methoxybenzoate 95%

Supplier: AMBEED, INC
Warning

A213487-1G A213487-250MG A213487-25G A213487-5G
77030-576EA 55 USD InStock
77030-576 77030-578 77030-580 77030-582
Methyl-4-(bromomethyl)-2-methoxybenzoate 95%
Methyl-4-(bromomethyl)-2-methoxybenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₁BrO₃
MDL Number: MFCD11976783
CAS Number: 74733-27-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty