Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-methylbenzotrifluoride 95%

Supplier: AMBEED, INC
Warning

A213657-100MG A213657-1G A213657-250MG A213657-5G
76999-680EA 56 USD InStock
76999-680 76999-682 76999-684 76999-686
2-Bromo-6-methylbenzotrifluoride 95%
2-Bromo-6-methylbenzotrifluoride
Precaución: for Research Use Only


Formula: C₈H₆BrF₃
MDL Number: MFCD01631583
CAS Number: 944268-56-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty