Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-N-methylnicotinamide 95%

Supplier: AMBEED, INC
Warning

A213933-10G A213933-1G A213933-250MG A213933-5G
76986-828EA 700 USD InStock
76986-828 76986-830 76986-832 76986-834
4-Amino-N-methylnicotinamide 95%
4-Amino-N-methylnicotinamide
Precaución: for Research Use Only


Formula: C₇H₉N₃O
MDL Number: MFCD18802374
CAS Number: 910656-00-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty