Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-Boc-2-(aminomethyl)pyrrolidine 97%

Supplier: AMBEED, INC
Warning

A215239-10G A215239-1G A215239-250MG A215239-25G A215239-5G
76999-756EA 49 USD InStock
76999-756 76999-758 76999-760 76999-762 76999-764
(S)-1-Boc-2-(aminomethyl)pyrrolidine 97%
(S)-1-Boc-2-(aminomethyl)pyrrolidine
Precaución: for Research Use Only


Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD03419257
CAS Number: 119020-01-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty