Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)imidazo[1,2-a]pyridine-8-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A215257-100MG A215257-1G A215257-250MG
77030-636EA 97 USD InStock
77030-636 77030-638 77030-640
2-(Trifluoromethyl)imidazo[1,2-a]pyridine-8-carboxylic acid 98%
2-(Trifluoromethyl)imidazo[1,2-a]pyridine-8-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₅F₃N₂O₂
MDL Number: MFCD11976231
CAS Number: 73221-20-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty