Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-2-(4-hydroxyphenyl)acetic acid 98% +

Supplier: AMBEED, INC
Warning

A216077-1G A216077-250MG A216077-25G A216077-5G
76999-788EA 6 USD InStock
76999-788 76999-790 76999-792 76999-794
2-Hydroxy-2-(4-hydroxyphenyl)acetic acid 98% +
2-Hydroxy-2-(4-hydroxyphenyl)acetic acid
Precaución: for Research Use Only


Formula: C₈H₈O₄
MW: 168.14 g/mol
MDL Number: MFCD00004234
CAS Number: 1198-84-1
Order Now
Specification Test Results
Loss on drying ≤ 10%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty