Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Diphenylphosphino)ethylammonium tetrafluoroborate 98%, Ambeed.Inc

Supplier: AMBEED, INC
Danger

A216348-1G A216348-250MG A216348-5G
76986-904EA 75 USD InStock
76986-904 77683-618 77683-619
2-(Diphenylphosphino)ethylammonium tetrafluoroborate 98%, Ambeed.Inc
2-(Diphenylphosphino)ethylammonium tetrafluoroborate
Precaución: For research use only.


Formula: C₁₄H₁₇BF₄NP
MDL Number: MFCD17014009
CAS Number: 1222630-32-1
UN: 3263
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty