Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-bromo-6-chloropyridine 97%

Supplier: AMBEED, INC
Warning

A217923-100MG A217923-250MG A217923-5G
77030-698EA 10 USD InStock
77030-698 77030-700 77667-820
2-Amino-3-bromo-6-chloropyridine 97%
2-Amino-3-bromo-6-chloropyridine
Precaución: for Research Use Only


Formula: C₅H₄BrClN₂
MW: 207.46 g/mol
MDL Number: MFCD13189343
CAS Number: 442127-50-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty