Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Isopropyl-1H-pyrazol-4-yl)methanol 98+%

Supplier: AMBEED, INC
Warning

A218974-100MG A218974-1G A218974-250MG
77030-732EA 12 USD InStock
77030-732 77030-734 77030-736
(1-Isopropyl-1H-pyrazol-4-yl)methanol 98+%
(1-Isopropyl-1H-pyrazol-4-yl)methanol
Precaución: for Research Use Only


Formula: C₇H₁₂N₂O
MW: 140.19 g/mol
MDL Number: MFCD08059963
CAS Number: 1007542-22-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty