Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxycyclohexanamine 96%

Supplier: AMBEED, INC
Danger

A220226-1G A220226-250MG A220226-5G
77030-778EA 65 USD InStock
77030-778 77030-780 77030-782
4-Methoxycyclohexanamine 96%
4-Methoxycyclohexanamine
Precaución: for Research Use Only


Formula: C₇H₁₅NO
MDL Number: MFCD17011799
CAS Number: 4342-46-5
UN: 2734
ADR: 8,3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty