Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Formyl-2-methylbenzonitrile 98%

Supplier: AMBEED, INC
Warning

A220370-100MG A220370-1G A220370-250MG
77030-788EA 30 USD InStock
77030-788 77030-790 77030-792
5-Formyl-2-methylbenzonitrile 98%
5-Formyl-2-methylbenzonitrile
Precaución: for Research Use Only


Formula: C₉H₇NO
MDL Number: MFCD12197728
CAS Number: 27613-36-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty