Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-homocyclohexylalanine 97%

Supplier: AMBEED, INC
Warning

A220545-100MG A220545-10G A220545-1G A220545-250MG A220545-5G
76999-892EA 6 USD InStock
76999-892 76999-894 76999-896 76999-898 76999-900
Fmoc-D-homocyclohexylalanine 97%
Fmoc-D-homocyclohexylalanine
Precaución: for Research Use Only


Formula: C₂₅H₂₉NO₄
MW: 407.51 g/mol
MDL Number: MFCD02179644
CAS Number: 269078-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty