Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-3-nitropicolinonitrile 97%

Supplier: AMBEED, INC
Warning

A220930-100MG A220930-1G A220930-250MG
76987-044EA 7 USD InStock
76987-044 76987-046 76987-048
5-Fluoro-3-nitropicolinonitrile 97%
5-Fluoro-3-nitropicolinonitrile
Precaución: for Research Use Only


Formula: C₆H₂FN₃O₂
MDL Number: MFCD11040666
CAS Number: 1033202-51-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty