Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(5-Chloro-2-methoxyphenyl)-3-hydroxy-2-naphthamide 98%

Supplier: AMBEED, INC
Danger

A221332-1G A221332-5G
77030-824EA 22 USD InStock
77030-824 77030-826
N-(5-Chloro-2-methoxyphenyl)-3-hydroxy-2-naphthamide 98%
N-(5-Chloro-2-methoxyphenyl)-3-hydroxy-2-naphthamide
Precaución: for Research Use Only


Formula: C₁₈H₁₄ClNO₃
MDL Number: MFCD00051171
CAS Number: 137-52-0
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty