Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Quinolineboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A221686-1G A221686-250MG A221686-5G
77030-828EA 16 USD InStock
77030-828 77030-830 77030-832
6-Quinolineboronic acid pinacol ester 98%
6-Quinolineboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₁₈BNO₂
MDL Number: MFCD06411327
CAS Number: 406463-06-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty