Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Diamino-2-fluorobenzotrifluoride 98%

Supplier: AMBEED, INC
Warning

A221742-10G A221742-1G A221742-250MG
76987-076EA 742 USD InStock
76987-076 76987-078 76987-080
4,5-Diamino-2-fluorobenzotrifluoride 98%
4,5-Diamino-2-fluorobenzotrifluoride
Precaución: for Research Use Only


Formula: C₇H₆F₄N₂
MDL Number: MFCD04038250
CAS Number: 179062-06-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty