Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-bromo-3-(trifluoromethyl)pyridine 98%

Supplier: AMBEED, INC
Warning

A221840-100G A221840-10G A221840-1G A221840-25G A221840-5G
77030-834EA 440 USD InStock
77030-834 77030-836 77030-838 77030-840 77030-842
2-Amino-5-bromo-3-(trifluoromethyl)pyridine 98%
2-Amino-5-bromo-3-(trifluoromethyl)pyridine
Precaución: for Research Use Only


Formula: C₆H₄BrF₃N₂
MDL Number: MFCD10758088
CAS Number: 79456-34-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty