Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-fluoro-6-methoxybenzoic acid 95%

Supplier: AMBEED, INC
Warning

A222806-100MG A222806-1G A222806-250MG
77013-298EA 102 USD InStock
77013-298 77013-300 77013-302
4-Chloro-2-fluoro-6-methoxybenzoic acid 95%
4-Chloro-2-fluoro-6-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₈H₆ClFO₃
MDL Number: MFCD11845551
CAS Number: 1082042-25-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty