Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2H-Chromene-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A222849-10G A222849-1G A222849-250MG A222849-5G
77013-304EA 256 USD InStock
77013-304 77013-306 77013-308 77013-310
2H-Chromene-3-carboxylic acid 98%
2H-Chromene-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₈O₃
MDL Number: MFCD00202020
CAS Number: 22649-28-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty