Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-oxoindoline-1-carboxylate 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A227583-1G A227583-250MG A227583-5G
76987-204EA 34 USD InStock
76987-204 76987-206 77683-660
tert-Butyl 2-oxoindoline-1-carboxylate 95%, Ambeed.Inc
tert-Butyl 2-oxoindoline-1-carboxylate
Precaución: For research use only.


Formula: C₁₃H₁₅NO₃
MW: 233.26 g/mol
MDL Number: MFCD09032873
CAS Number: 214610-10-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty