Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminopyridine-4-boronic acid pinacol ester 96%

Supplier: AMBEED, INC
Danger

A228108-100MG A228108-1G A228108-250MG
77030-972EA 22 USD InStock
77030-972 77030-974 77030-976
2-Aminopyridine-4-boronic acid pinacol ester 96%
2-Aminopyridine-4-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₁H₁₇BN₂O₂
MDL Number: MFCD09607735
CAS Number: 1195995-72-2
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty