Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A228783-100MG A228783-250MG A228783-5G
77030-988EA 12 USD InStock
77030-988 77030-990 77030-992
3-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester 95%
3-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₂₁BO₄
MDL Number: MFCD13190786
CAS Number: 955929-54-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty