Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,6-dichlorobenzothiazole 95%

Supplier: AMBEED, INC
Warning

A228997-10G A228997-1G A228997-250MG A228997-5G
77000-136EA 68 USD InStock
77000-136 77000-138 77000-140 77000-142
2-Amino-4,6-dichlorobenzothiazole 95%
2-Amino-4,6-dichlorobenzothiazole
Precaución: for Research Use Only


Formula: C₇H₄Cl₂N₂S
MDL Number: MFCD01123002
CAS Number: 16582-59-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty