Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-(4-bromophenyl)acetate 97%

Supplier: AMBEED, INC
Warning

A229160-100MG A229160-1G
76987-272EA 36 USD InStock
76987-272 76987-274
tert-Butyl 2-(4-bromophenyl)acetate 97%
tert-Butyl 2-(4-bromophenyl)acetate
Precaución: for Research Use Only


Formula: C₁₂H₁₅BrO₂
MW: 271.15 g/mol
MDL Number: MFCD11975535
CAS Number: 33155-58-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty