Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-(N-(Thiazol-2-yl)sulfamoyl)phenyl)acetamide 95%

Supplier: AMBEED, INC
Warning

A229669-100MG A229669-1G A229669-250MG
76987-298EA 47 USD InStock
76987-298 76987-300 76987-302
N-(4-(N-(Thiazol-2-yl)sulfamoyl)phenyl)acetamide 95%
N-(4-(N-(Thiazol-2-yl)sulfamoyl)phenyl)acetamide


Formula: C₁₁H₁₁N₃O₃S₂
MDL Number: MFCD00248974
CAS Number: 127-76-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty