Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyridine 95%

Supplier: AMBEED, INC
Warning

A231370-1G A231370-250MG A231370-5G
76987-402EA 40 USD InStock
76987-402 76987-404 76987-406
8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyridine 95%
8-Chloro-3-methyl[1,2,4]triazolo[4,3-a]pyridine
Precaución: for Research Use Only


Formula: C₇H₆ClN₃
MW: 167.6 g/mol
MDL Number: MFCD09258771
CAS Number: 929000-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty