Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-4-pyridineboronic acid 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A232342-100MG A232342-5G A232342-1G
76987-434EA 14 USD InStock
76987-434 76987-436 77683-676
3-Methyl-4-pyridineboronic acid 97%, Ambeed.Inc
3-Methyl-4-pyridineboronic acid
Precaución: For research use only.


Formula: C₆H₈BNO₂
MW: 136.9400 g/mol
MDL Number: MFCD03095365
CAS Number: 894808-72-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty