Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-chloropyrazolo[1,5-a]pyrimidine-3-carboxylate 97%

Supplier: AMBEED, INC

A232728-100MG A232728-1G A232728-250MG A232728-5G
76987-440EA 22 USD InStock
76987-440 76987-442 76987-444 76987-446
Methyl 5-chloropyrazolo[1,5-a]pyrimidine-3-carboxylate 97%
Methyl 5-chloropyrazolo[1,5-a]pyrimidine-3-carboxylate
Precaución: for Research Use Only


Formula: C₈H₆ClN₃O₂
MDL Number: MFCD22551533
CAS Number: 1224944-51-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty