Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-amino-4-(methylamino)benzenecarboxylate 96%

Supplier: AMBEED, INC
Warning

A232745-10G A232745-1G A232745-250MG
77000-244EA 320 USD InStock
77000-244 77000-246 77000-248
Methyl-3-amino-4-(methylamino)benzenecarboxylate 96%
Methyl-3-amino-4-(methylamino)benzenecarboxylate
Precaución: for Research Use Only


Formula: C₉H₁₂N₂O₂
MDL Number: MFCD12922710
CAS Number: 66315-16-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty