Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid 95%

Supplier: AMBEED, INC
Warning

A233391-100MG A233391-1G A233391-250MG
77013-570EA 11 USD InStock
77013-570 77013-572 77013-574
4-(2-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid 95%
4-(2-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₅H₁₆BClO₃
MW: 290.5500 g/mol
MDL Number: MFCD06411361
CAS Number: 849052-15-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty