Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl 2-oxaspiro[3.3]heptane-6,6-dicarboxylate 97%

Supplier: AMBEED, INC
Warning

A233593-100MG A233593-1G
76987-456EA 126 USD InStock
76987-456 76987-458
Diethyl 2-oxaspiro[3.3]heptane-6,6-dicarboxylate 97%
Diethyl 2-oxaspiro[3.3]heptane-6,6-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₈O₅
MW: 242.26 g/mol
MDL Number: MFCD18909536
CAS Number: 26593-41-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty