Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di(pyridin-2-yl)methanol 95%

Supplier: AMBEED, INC
Warning

A235531-1G A235531-250MG A235531-5G
76987-498EA 108 USD InStock
76987-498 76987-500 76987-502
Di(pyridin-2-yl)methanol 95%
Di(pyridin-2-yl)methanol
Precaución: for Research Use Only


Formula: C₁₁H₁₀N₂O
MDL Number: MFCD09838260
CAS Number: 35047-29-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty