Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-3-fluoro-2-nitrophenol 95%

Supplier: AMBEED, INC
Danger

A238418-100MG A238418-1G A238418-250MG A238418-5G
76987-558EA 40 USD InStock
76987-558 76987-560 76987-562 76987-564
6-Bromo-3-fluoro-2-nitrophenol 95%
6-Bromo-3-fluoro-2-nitrophenol
Precaución: for Research Use Only


Formula: C₆H₃BrFNO₃
MW: 235.995 g/mol
MDL Number: MFCD23701729
CAS Number: 872363-59-2
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty