Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-Amino-4,4-diphenylbutanoic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A238657-100MG A238657-250MG
76987-566EA 39 USD InStock
76987-566 76987-568
(S)-3-Amino-4,4-diphenylbutanoic acid hydrochloride 95%
(S)-3-Amino-4,4-diphenylbutanoic acid hydrochloride
Precaución: for Research Use Only


Formula: C₁₆H₁₈ClNO₂
MDL Number: MFCD03092955
CAS Number: 332062-01-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty