Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2,9-diazaspiro[5.5]undecane-9-carboxylate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A238729-100MG A238729-1G A238729-250MG
76987-570EA 6 USD InStock
76987-570 76987-572 76987-574
tert-Butyl 2,9-diazaspiro[5.5]undecane-9-carboxylate hydrochloride 95%
tert-Butyl 2,9-diazaspiro[5.5]undecane-9-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₁₄H₂₇ClN₂O₂
MDL Number: MFCD22209368
CAS Number: 1279866-58-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty