Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl ((3-hydroxycyclobutyl)methyl)carbamate 95%

Supplier: AMBEED, INC
Warning

A239354-100MG A239354-1G A239354-250MG
76987-584EA 23 USD InStock
76987-584 76987-586 76987-588
tert-Butyl ((3-hydroxycyclobutyl)methyl)carbamate 95%
tert-Butyl ((3-hydroxycyclobutyl)methyl)carbamate
Precaución: for Research Use Only


Formula: C₁₀H₁₉NO₃
MDL Number: MFCD13659430
CAS Number: 167081-41-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty