Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3'-Bicarbazole 95%

Supplier: AMBEED, INC
Warning

A240694-100G A240694-1G A240694-25G A240694-5G
76987-616EA 222 USD InStock
76987-616 76987-618 76987-620 76987-622
3,3'-Bicarbazole 95%
3,3'-Bicarbazole
Precaución: for Research Use Only


Formula: C₂₄H₁₆N₂
MDL Number: MFCD11046470
CAS Number: 1984-49-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty