Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Formylpyridine-3-boronic acid, pinacol ester 98%

Supplier: AMBEED, INC
Warning

A241606-100MG A241606-250MG
76987-634EA 37 USD InStock
76987-634 76987-636
6-Formylpyridine-3-boronic acid, pinacol ester 98%
6-Formylpyridine-3-boronic acid, pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₆BNO₃
MDL Number: MFCD06659508
CAS Number: 1073354-14-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty