Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-methoxypyridine 98%

Supplier: AMBEED, INC
Danger

A242545-10G A242545-1G A242545-25G A242545-5G
77000-462EA 12 USD InStock
77000-462 77000-464 77000-466 77000-468
2-Fluoro-6-methoxypyridine 98%
2-Fluoro-6-methoxypyridine
Precaución: for Research Use Only


Formula: C₆H₆FNO
MDL Number: MFCD09998161
CAS Number: 116241-61-3
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty