Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Pyridin-4-yl)benzoic acid methyl ester 97%

Supplier: AMBEED, INC
Warning

A243315-100MG A243315-1G A243315-250MG A243315-5G
77013-808EA 12 USD InStock
77013-808 77013-810 77013-812 77013-814
4-(Pyridin-4-yl)benzoic acid methyl ester 97%
4-(Pyridin-4-yl)benzoic acid methyl ester
Precaución: for Research Use Only


Formula: C₁₃H₁₁NO₂
MDL Number: MFCD04114575
CAS Number: 106047-17-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty