Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-indoleboronic acid pinacol ester 96%

Supplier: AMBEED, INC
Warning

A243568-1G A243568-250MG A243568-5G
77013-816EA 167 USD InStock
77013-816 77013-818 77013-820
5-Bromo-2-indoleboronic acid pinacol ester 96%
5-Bromo-2-indoleboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₁₇BBrNO₂
MDL Number: MFCD11858360
CAS Number: 1256358-92-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty