Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyanopyrazine 98%

Supplier: AMBEED, INC
Warning

A244035-1000G A244035-100G A244035-25G A244035-500G A244035-5G
76987-684EA 138 USD InStock
76987-684 76987-686 76987-688 76987-690 76987-692 76987-694
2-Cyanopyrazine 98%
2-Cyanopyrazine
Precaución: for Research Use Only


Formula: C₅H₃N₃
MDL Number: MFCD00049361
CAS Number: 19847-12-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty