Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydro-4H-pyran-4-one 98%

Supplier: AMBEED, INC
Danger

A245086-10G A245086-25G
77031-342EA 8 USD InStock
77031-342 77031-344 77031-346 77031-348
Tetrahydro-4H-pyran-4-one 98%
Tetrahydro-4H-pyran-4-one
Precaución: for Research Use Only


Formula: C₅H₈O₂
MDL Number: MFCD00006581
CAS Number: 29943-42-8
UN: 1224
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty