Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-chloro-2-hydroxybenzoate 98%

Supplier: AMBEED, INC
Warning

A247144-1G A247144-1MG A247144-250MG A247144-5G
77031-386EA 24 USD InStock
77031-386 77031-388 77031-390 77031-392
Methyl-3-chloro-2-hydroxybenzoate 98%
Methyl-3-chloro-2-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₈H₇ClO₃
MW: 186.5924 g/mol
MDL Number: MFCD00128116
CAS Number: 52159-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty