Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl imidazo[1,5-a]pyridine-1-carboxylate 98%

Supplier: AMBEED, INC
Warning

A248323-1G A248323-250MG A248323-5G
77013-932EA 74 USD InStock
77013-932 77013-934 77013-936
Ethyl imidazo[1,5-a]pyridine-1-carboxylate 98%
Ethyl imidazo[1,5-a]pyridine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂O₂
MW: 190.1986 g/mol
MDL Number: MFCD01237556
CAS Number: 119448-87-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty